ADVH7F383787 AChemBlock

2-amino-2-(2,4-difluorophenyl)ethan-1-ol

  • MDL number MFCD11213676
  • Cas Number : 872574-40-8
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/2-amino-2-2-4-difluorophenyl-ethan-1-ol.html

Safety Information

signalword Warning
pcodes P261; P305 + P351 + P338
MSDS https://www.achemblock.com/ac_sds?Catalog=S88049
pictograms GHS07
hstatements H302; H315; H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart