ADVH7F3838D1 AChemBlock

4,4'-dibromo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol

  • MDL number MFCD16660947
  • Cas Number : 681481-91-4
  • Molecular Weight : 410.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at 0-8 �C
Shipping Temperature Store at 0-8 �C
Country of Origin CN
Product Link https://www.achemblock.com/4-4-dibromo-2-2-3-3-tetrahydro-1-1-spirobi-indene-7-7-diol.html

Safety Information

signalword Danger
pcodes P261; P264; P270; P272; P280; P301+PP310; P302+P352; P305+P351+P338; P313+P333; P337+P313; P330; P363; P405; P501
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88613
pictograms GHS06
hstatements H301; H317; H319
Availability details for
Quantity
  • Availability
  • Add to Cart