ADVH7F3838DB AChemBlock

4,4'-(4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine-2,6-diyl)bis(naphthalen-1(5H)-one)

  • MDL number MFCD32173555
  • Cas Number : 929097-93-8
  • Molecular Weight : 616.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/4-4-4-hydroxydinaphtho-2-1-d-1-2-f-1-3-2-dioxaphosphepine-2-6-diyl-bis-naphthalen-1-5h-one.html

Safety Information

signalword Warning
pcodes P264; P280; P302+P352; P305+P351+P338; P332+P313; P337+P313; P362
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88626
pictograms GHS07
hstatements H315; H319; H320
Availability details for
Quantity
  • Availability
  • Add to Cart