ADVH7F3838E2 AChemBlock

tert-butyl 4-(2,4-difluorobenzoyl)piperidine-1-carboxylate

  • MDL number MFCD12068396
  • Cas Number : 1159825-99-6
  • Molecular Weight : 325.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/tert-butyl-4-2-4-difluorobenzoyl-piperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P280; P305 + P351 + P338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88696
pictograms GHS07
hstatements H317; H319
Availability details for
Quantity
  • Availability
  • Add to Cart