ADVH7F383977 AChemBlock

4-amino-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD08234783
  • Cas Number : 155815-95-5
  • Molecular Weight : 126.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin CN
Product Link https://www.achemblock.com/4-amino-1h-pyrrole-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P280; P305 + P351 + P338
MSDS https://www.achemblock.com/ac_sds?Catalog=S89137
pictograms GHS07
hstatements H302; H317; H319
Availability details for
Quantity
  • Availability
  • Add to Cart