ADVH9454686A AChemBlock

bicyclo[1.1.1]pentane-1-carbonitrile

  • MDL number MFCD29037333
  • Cas Number : 126332-44-3
  • Molecular Weight : 93.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at 0-8 �C
Shipping Temperature Store at 0-8 �C
Country of Origin CN
Product Link https://www.achemblock.com/bicyclo-1-1-1-pentane-1-carbonitrile.html

Safety Information

signalword Danger
pcodes P201;P202;P210;P233;P240;P301+330+331;P303+361+353
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=P48972
pictograms GHS02;GHS05;GHS06;GHS08;GHS09
hstatements H222;H302;H315;H331;H341;H351;H373;H411
Availability details for
Quantity
  • Availability
  • Add to Cart