ADVH94546AA3 AChemBlock

methyl (1S,3R)-rel-3-(aminomethyl)cyclohexanecarboxylate hydrochloride

  • MDL number MFCD31928787
  • Cas Number : 916211-60-4
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/methyl-1s-3r-rel-3-aminomethyl-cyclohexanecarboxylate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261; P305 + P351 + P338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S73062
pictograms GHS07
hstatements H302; H315; H319; H335
Availability details for
Quantity
  • Availability
  • Add to Cart