ADVH94546BEE AChemBlock

6-bromo-5-fluoro-1H-indole-3-carbaldehyde

  • MDL number MFCD16609950
  • Cas Number : 1227496-42-5
  • Molecular Weight : 242.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Store at Room Tem.
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/6-bromo-5-fluoro-1h-indole-3-carbaldehyde.html

Safety Information

signalword Warning
pcodes P280; P305 + P351 + P338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82160
pictograms GHS07
hstatements H317; H319
Availability details for
Quantity
  • Availability
  • Add to Cart