AMBH2D6EBA23 Ambeed, Inc.

Dabrafenib Mesylate

  • MDL number MFCD20922872
  • Cas Number : 1195768-06-9
  • Molecular Weight : 615.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1195768-06-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1195768-06-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart