AMBH2D6EBAA8 Ambeed, Inc.

Verubecestat

  • MDL number MFCD28963974
  • Cas Number : 2095432-65-6
  • Molecular Weight : 523.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (R)-5-Fluoro-N-(4-fluoro-3-(3-imino-2,5-dimethyl-1,1-dioxido-1,2,4-thiadiazinan-5-yl)phenyl)picolinamide 2,2,2-trifluoroacetate
Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/2095432-65-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2095432-65-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart