AMBH2D6EBB18 Ambeed, Inc.

6-Chloro-5-(2-chloroethyl)indolin-2-one

  • MDL number MFCD03411598
  • Cas Number : 118289-55-7
  • Molecular Weight : 230.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/118289-55-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/118289-55-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart