AMBH2D6EBB89 Ambeed, Inc.

2-Amino-5-bromo-4,6-dimethylpyrimidine

  • MDL number MFCD00160498
  • Cas Number : 4214-57-7
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/4214-57-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/4214-57-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart