AMBH2D6EBFD6 Ambeed, Inc.

5-Chloro-2,4-difluorobenzonitrile

  • MDL number MFCD06659465
  • Cas Number : 146780-26-9
  • Molecular Weight : 173.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-51
Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 205�t 760 mmHg
Product Link https://www.ambeed.com/products/146780-26-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/146780-26-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart