AMBH2D6EC4A7 Ambeed, Inc.

5-Bromo-4-chloro-2-fluorobenzoic acid

  • MDL number MFCD00672928
  • Cas Number : 289038-22-8
  • Molecular Weight : 253.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-141°C
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 329.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/289038-22-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/289038-22-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart