AMBH2D6EC4D7 Ambeed, Inc.

(5-Chloro-1H-indol-3-yl)methanamine

  • MDL number MFCD06657113
  • Cas Number : 113188-83-3
  • Molecular Weight : 180.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 177.4
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 369.8�t 760 mmHg
Product Link https://www.ambeed.com/products/113188-83-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/113188-83-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart