AMBH2D6EC4F6 Ambeed, Inc.

5-Chloro-1H-indole-3-carbonitrile

  • MDL number MFCD07776618
  • Cas Number : 194490-14-7
  • Molecular Weight : 176.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/194490-14-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/194490-14-7.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart