AMBH2D6EC50F Ambeed, Inc.

3-Fluoro-2-(trifluoromethyl)aniline

  • MDL number MFCD06658260
  • Cas Number : 123973-22-8
  • Molecular Weight : 179.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/123973-22-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/123973-22-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart