AMBH2D6EC517 Ambeed, Inc.

3-Fluoro-4-(trifluoromethyl)aniline

  • MDL number MFCD00190120
  • Cas Number : 69411-68-3
  • Molecular Weight : 179.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/69411-68-3.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P312
MSDS https://www.ambeed.com/products/69411-68-3.html
pictograms GHS06
hstatements H302-H311-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart