AMBH2D6EC536 Ambeed, Inc.

5-Ethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

  • MDL number MFCD00079592
  • Cas Number : 34171-37-4
  • Molecular Weight : 156.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-193°C
Purity 97
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/34171-37-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/34171-37-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart