AMBH2D6EC5B6 Ambeed, Inc.

5-Methyl-2-(trifluoromethyl)aniline

  • MDL number MFCD04972928
  • Cas Number : 106877-29-6
  • Molecular Weight : 175.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/106877-29-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/106877-29-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart