AMBH2D6EC5BB Ambeed, Inc.

5-Methyl-2-pentylphenol

  • MDL number MFCD00127710
  • Cas Number : 1300-94-3
  • Molecular Weight : 178.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1300-94-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1300-94-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart