AMBH2D6EC9A1 Ambeed, Inc.

H-D-Glu(OMe)-OH

  • MDL number MFCD00237069
  • Cas Number : 6461-04-7
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176°C
Synonym (R)-2-Amino-5-methoxy-5-oxopentanoic acid
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 316.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/6461-04-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6461-04-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart