AMBH2D6ECA8C Ambeed, Inc.

Ethyl 2-(tetrahydrofuran-2-yl)acetate

  • MDL number MFCD00059735
  • Cas Number : 2434-02-8
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 89.3
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 206.1�t 760 mmHg
Product Link https://www.ambeed.com/products/2434-02-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2434-02-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart