AMBH2D6ECAC8 Ambeed, Inc.

4-Bromo-2-furoic acid

  • MDL number MFCD00092494
  • Cas Number : 3439-02-9
  • Molecular Weight : 190.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3439-02-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/3439-02-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart