AMBH2D6ECB75 Ambeed, Inc.

2-Chloro-4-methylpyrimidine-5-carboxylic acid

  • MDL number MFCD09863167
  • Cas Number : 188781-10-4
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/188781-10-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/188781-10-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart