AMBH2D6ECC57 Ambeed, Inc.

tert-Butyl 3,6-diazabicyclo[3.2.0]heptane-6-carboxylate

  • MDL number MFCD09991605
  • Cas Number : 122848-57-1
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/122848-57-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/122848-57-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart