AMBH2D6ECCDF Ambeed, Inc.

6-Bromo-4-methoxyquinoline

  • MDL number MFCD04036192
  • Cas Number : 874792-20-8
  • Molecular Weight : 238.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/874792-20-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/874792-20-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart