AMBH2D6ECD07 Ambeed, Inc.

6-Chloro-3-iodo-2-methylpyridine

  • MDL number MFCD09037461
  • Cas Number : 249291-79-0
  • Molecular Weight : 253.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-33°C
Purity 97
Flash Point 111.3°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 260.4°C at 760 mmHg
Product Link https://www.ambeed.com/products/249291-79-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/249291-79-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart