AMBH2D6ECD52 Ambeed, Inc.

6-Chloro-3-methylimidazo[1,2-b]pyridazine

  • MDL number MFCD09991804
  • Cas Number : 137384-48-6
  • Molecular Weight : 167.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/137384-48-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/137384-48-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart