AMBH2D6ECD58 Ambeed, Inc.

6-Chloro-3-hydroxypyridazine-4-carboxylic acid

  • MDL number MFCD12756225
  • Cas Number : 50681-26-0
  • Molecular Weight : 174.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/50681-26-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/50681-26-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart