AMBH2D6ECD5E Ambeed, Inc.

6-Chloro-4-methoxypicolinonitrile

  • MDL number MFCD03844440
  • Cas Number : 193074-46-3
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 121.6
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 277.5�0�t 760 mmHg
Product Link https://www.ambeed.com/products/193074-46-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/193074-46-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart