AMBH2D6ECE1B Ambeed, Inc.

6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one

  • MDL number MFCD00042479
  • Cas Number : 22123-19-9
  • Molecular Weight : 177.1200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/22123-19-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/22123-19-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart