AMBH2D6ECFDF Ambeed, Inc.

9,9'-Bianthracene

  • MDL number MFCD00130216
  • Cas Number : 1055-23-8
  • Molecular Weight : 354.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1055-23-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1055-23-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart