AMBH2D6ED060 Ambeed, Inc.

H-Phe(4-NO2)-OH

  • MDL number MFCD00051221
  • Cas Number : 949-99-5
  • Molecular Weight : 210.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-239
Synonym (S)-2-Amino-3-(4-nitrophenyl)propanoic acid
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 414.1�t 760 mmHg
Product Link https://www.ambeed.com/products/949-99-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/949-99-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart