AMBH2D6ED087 Ambeed, Inc.

Alizapride Hydrochloride

  • MDL number MFCD02183928
  • Cas Number : 59338-87-3
  • Molecular Weight : 351.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/59338-87-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59338-87-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart