AMBH2D6ED0FA Ambeed, Inc.

Amezinium methylsulfate

  • MDL number MFCD00865972
  • Cas Number : 30578-37-1
  • Molecular Weight : 313.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/30578-37-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/30578-37-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart