AMBH2D6ED11B Ambeed, Inc.

4-Aminobenzene-1,3-disulfonic acid

  • MDL number MFCD00035903
  • Cas Number : 137-51-9
  • Molecular Weight : 253.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97102
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/137-51-9.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/137-51-9.html
pictograms GHS05,GHS07
hstatements H302-H314
Availability details for
Quantity
  • Availability
  • Add to Cart