AMBH2D6ED356 Ambeed, Inc.

(S)-1-(2,4-Difluorophenyl)ethanamine

  • MDL number MFCD06761834
  • Cas Number : 845252-02-0
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 72.8
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 175.4�t 760 mmHg
Product Link https://www.ambeed.com/products/845252-02-0.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/845252-02-0.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart