AMBH2D6ED9B6 Ambeed, Inc.

2-Benzamido-3-methylbutanoic acid

  • MDL number MFCD00038283
  • Cas Number : 2901-80-6
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 132
bp 451.8�t 760 mmHg
Iupac Name N-benzoylvaline
Inchi Code 1S/C12H15NO3/c1-8(2)10(12(15)16)13-11(14)9-6-4-3-5-7-9/h3-8,10H,1-2H3,(H,13,14)(H,15,16)
Inchi Key MIYQNOPLWKCHED-UHFFFAOYSA-N

Description

Melting Point 132
Purity 95%
Flash Point 227
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 451.8�t 760 mmHg
Product Link https://www.ambeed.com/products/2901-80-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/2901-80-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart