AMBH2D6ED9CB Ambeed, Inc.

Benzyl(phenyl)sulfane

  • MDL number MFCD00003066
  • Cas Number : 831-91-4
  • Molecular Weight : 200.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 40-44
bp 321.1�t 760 mmHg
Iupac Name [(phenylsulfanyl)methyl]benzene
Inchi Code 1S/C13H12S/c1-3-7-12(8-4-1)11-14-13-9-5-2-6-10-13/h1-10H,11H2
Inchi Key LKMCJXXOBRCATQ-UHFFFAOYSA-N

Description

Melting Point 40-44
Purity 98%
Flash Point 113
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 321.1�t 760 mmHg
Product Link https://www.ambeed.com/products/831-91-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/831-91-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart