AMBH2D6EDA21 Ambeed, Inc.

Sodium 1,4-dioxo-1,4-bis(tridecyloxy)butane-2-sulfonate

  • MDL number MFCD00047784
  • Cas Number : 2673-22-5
  • Molecular Weight : 584.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 65
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2673-22-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2673-22-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart