AMBH2D6EDA25 Ambeed, Inc.

1,1,1-Triphenyl-N-(triphenylphosphoranylidene)phosphoraniminium chloride

  • MDL number MFCD00151523
  • Cas Number : 21050-13-5
  • Molecular Weight : 574.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/21050-13-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/21050-13-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart