AMBH2D6EDA28 Ambeed, Inc.

Sulbutiamine

  • MDL number MFCD01726427
  • Cas Number : 3286-46-2
  • Molecular Weight : 702.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Disulfanediylbis(4-(N-((4-amino-2-methylpyrimidin-5-yl)methyl)formamido)pent-3-ene-3,1-diyl) bis(2-methylpropanoate)
Purity 98
Flash Point 493.5
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 892.5�t 760 mmHg
Product Link https://www.ambeed.com/products/3286-46-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3286-46-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart