AMBH2D6EDA78 Ambeed, Inc.

1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethanone

  • MDL number MFCD00665892
  • Cas Number : 29096-60-4
  • Molecular Weight : 174.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/29096-60-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/29096-60-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart