AMBH2D6EDB25 Ambeed, Inc.

Calcium Levofolinate

  • MDL number MFCD08704249
  • Cas Number : 80433-71-2
  • Molecular Weight : 511.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Calcium 2-(4-(((2-amino-5-formyl-4-oxo-3,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioate
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/80433-71-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/80433-71-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart