AMBH2D6EDB49 Ambeed, Inc.

Calcium 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

  • MDL number MFCD00039119
  • Cas Number : 22454-86-0
  • Molecular Weight : 350.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/22454-86-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/22454-86-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart