AMBH2D6EDB73 Ambeed, Inc.

1-Cyano-4-(dimethylamino)pyridin-1-ium tetrafluoroborate

  • MDL number MFCD00011998
  • Cas Number : 59016-56-7
  • Molecular Weight : 234.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-200
Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/59016-56-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59016-56-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart