AMBH2D6EDBD6 Ambeed, Inc.

Chelerythrine Chloride

  • MDL number MFCD00060717
  • Cas Number : 3895-92-9
  • Molecular Weight : 383.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/3895-92-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/3895-92-9.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart