AMBH2D6EDBD7 Ambeed, Inc.

4-Methoxy-2-methylpyridine

  • MDL number MFCD06253892
  • Cas Number : 24103-75-1
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/24103-75-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/24103-75-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart