AMBH2D6EDC7E Ambeed, Inc.

Disodium Cromoglycate

  • MDL number MFCD22575281
  • Cas Number : 15826-37-6
  • Molecular Weight : 512.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/15826-37-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/15826-37-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart